missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Indoor/Outdoor BMP™71 Vinyl Labeling Tape

For use with BMP 71 Label Maker

Brand:  Brady™ 142348

 View more versions of this product

Product Code. 11785603

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Aggressive permanent adhesive sticks well to pipes, walls, equipment, windows and most clean, dry surfaces
  • Pliable—conforms well to irregular, curved or rough surfaces
  • Excellent resistance to oils, detergents, dirt, grime and many solvents
  • Five-year average outdoor durability in temperatures from 180° to -40° F (82° to -40°C)
  • Can be applied in temperatures as low as 0°F (-18°C)

Order Info

Each roll is 50 ft. long (15.2m)

Specifications

Specifications

BS5609:1986 Section 2 and section 3
Acrylic, Permanent
Gloss
Vinyl
Indoor and outdoor use
50 Gardner Units
5 Years
-40 to 180°F
0.004 in.
50 ft.
1 in.
BMP71 Label Printer Tapes
Orange
Tape
Labeling Tape
BMP™71 Label Maker
1
1 Roll
-40°C to +82°C
0.102 mm
15.24 m
25.40 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.