Learn More
Brady™ Label Maker Cartridge: Polyester, StainerBondz™
Label Maker Cartridge: StainerBondz™ Super Chemical Resistant Polyester (B-481) is designed for use in the slide staining process and other laboratory ID which requires superior chemical resistance.
Brand:  Brady™ 143302
 View more versions of this product
Product Code. 17861495
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • StainerBondz™ B-481 matte white polyester has a permanent acrylic adhesive and is thermal transfer printable with B30 series printers from Brady™
  • B-481 performs well in the commonly used H & E (hematoxilin and eosin) slide staining process
  • Excellent resistance to a wide variety of chemicals, oils and water
  • All-in-one cartridge includes label supply and R6400 Series ribbon
Specifications
Specifications
RoHS, ASTM D
Black
Polyester
Slide staining process and other laboratory identification which requires superior chemical resistance
1 Ea.
-80 to -3°C
0.0033 in.
0.9 in.
0.9 in.
Acrylic
Matte White
Non-sterile
BMP53 Label Printer
-112 to 266°F
40 lb./in.
0.9 x 0.9 in.
22.86 mm
12.70 mm