Learn More
Brady™ Label Maker Vinyl Cartridges (B-427) for BMP51, BMP53, BMP41 Printers
Designed to adhere to highly curved surfaces, wires and cables
Brand:  Brady™ 131573
 View more versions of this product
Product Code. 15153339
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Feature printable zone in white or yellow
  • Transparent, non printable area wraps around wires and cables for laminating printed texts
  • Offers resistance to abrasion, handling, solvents, water, oil and dirt
  • Available in various sizes
Specifications
Specifications
Black, Yellow
Label Maker Cartridge
1 Ea. (180 labels)
25.4 x 38.1 mm
25.4 mm
38.1 mm
Self-laminating Vinyl (B-427)
BMP™51,BMP™53,BMP™41 Printers
1 x 1.5 in.
1 in.
1.5 in.