missing translation for 'onlineSavingsMsg'
Learn More

Brady™ LABXPERT™ High-Performance Lab Polyester Labels

LABXPERT™ High-Performance Lab Polyester Labels are made of self-laminating polyester (B-461), a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.

65.97 € - 134.00 €

Specifications

Adhesive Type Acrylic, Permanent
Finish Matte
Format Label
Material Polyester
Product Type Label
View More Specs

Products 12
Product Code Brand Color Quantity Height (Metric) Description Price Quantity & Availability  
Product Code Brand Color Quantity Height (Metric) Description Price Quantity & Availability  
10512213
View Documents
Brady™
060943
Black, White, Translucent 150/Pk. 66.67 mm Long tail This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10225972
View Documents
Brady™
060932
Black, White, Translucent 450/Pk. 0.47 cm Long tail This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10694723
View Documents
Brady™
060933
Black, White, Translucent 100/Roll 9.52 cm Long tail This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10012002
View Documents
Brady™
060935
Black, White, Translucent 450/Pk. 0.95 cm Short tail This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12849998
View Documents
Brady™
060940
Black, White, Translucent 225/Pk. 1.27 cm Long tail This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10694533
View Documents
Brady™
060938
Black, White, Translucent 225/Pk. 1.52 cm Short tail This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10787784
View Documents
Brady™
060942
Black, White, Translucent 175/Pk. 1.27 cm Long tail This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10473552
View Documents
Brady™
060930
Black, White 1 Roll 0.95 cm Surface Mount This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10797784
View Documents
Brady™
060927
Black, White 1 Roll 2.54 cm Surface Mount This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10032012
View Documents
Brady™
060928
Black, White 1 Roll 3.17 cm Surface Mount This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10192892
View Documents
Brady™
060931
Black, White 1 Roll 1.27 cm Surface Mount This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10081982
View Documents
Brady™
060934
Black, White, Translucent 450/Pk. 12.70 mm Surface Mount This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Self-Laminating Polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • All-in-one cartridge includes ribbon and label supply
Specifications

Specifications

Acrylic, Permanent
Label
Label
-320 to 250°F
0.0019 in.
Matte
Polyester
Non-sterile
-196°C to +121°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.