Learn More
Brady™ LABXPERT™ Printer Labels
Specialty laboratory labels designed for use with these specific label printers
80.90 € - 112.00 €
Specifications
Description Surface Mount
Adhesive Type Acrylic, Permanent
Finish Matte
Product Type Laboratory Label
For Use With (Equipment) LabXpert™ Printer
View More Specs
Products 3
Product Code Brand Quantity Height (Metric) Material Width (English) Price Quantity & Availability  
Product Code Brand Quantity Height (Metric) Material Width (English) Price Quantity & Availability  
10153042
View Documents
Brady™
060905
175/Pk. 5.58 cm Freezerbondz I Polyester 1.5 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10522213
View Documents
Brady™
060939
175/Pk. 5.71 cm Self-Laminating Polyester 1.5 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10307132
View Documents
Brady™
060944
150/Pk. 1.52 cm Self-Laminating Polyester 2.62 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Printable, self-laminating high-performance laboratory identification for LABXPERT printer.
  • Polyester label withstands temps -196° to +121°C
Ultrathin construction prevents jamming in racks & centrifuge
Specifications
Specifications
Surface Mount
Matte
LabXpert™ Printer
Acrylic, Permanent
Laboratory Label