Learn More
Brady™ Nylon Cloth Laboratory Labels
Nylon Cloth Laboratory Labels (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
Brand:  Brady™ 143017
 View more versions of this product
Product Code. 13533310
  • 205.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Matte white polyamide coated nylon cloth material has a permanent acrylic adhesive and is thermal transfer printable
  • Recommended for indoor use
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
  • Service temperature of -40 to 193° F (-40 to 90° C)
Specifications
Specifications
RoHS
Acrylic, Permanent
Matte
Laboratory Label
Wiremarking and general labeling applications
27.94 mm
1500/Pk.
-40 to 193°F
80 lb./in.
0.165 mm
0.375 x 1 in.
25.4 mm
9 mm
Nylon Cloth Labels for the BBP33 Printer
White
Nylon Cloth
Non-sterile
BBP™33
2
LabelMark, MarkWare
-40°C to +90°C
0.0065 in.
12.07 mm
1 in.
0.3 in.