missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Nylon Cloth Laboratory Labels

Nylon Cloth Laboratory Labels (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.

Brand:  Brady™ 143049

182.40 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 15706879

  • 197.00 € / Pack of 1500
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Matte white polyamide coated nylon cloth material has a permanent acrylic adhesive and is thermal transfer printable
  • Recommended for indoor use
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
  • Service temperature of -40 to 193° F (-40 to 90° C)
Specifications

Specifications

RoHS
White
Nylon Cloth
Non-sterile
BBP™33
-40 to 193°F
80 lb./in.
0.25 x 2 in.
50.8 mm
6 mm
Acrylic, Permanent
Matte
Laboratory Label
Wiremarking and general labeling applications
1500/Pk.
-40°C to +90°C
0.0065 in.
2 in.
0.25 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers