Learn More
Brady™ Self-laminating Label Cartridge for BMP™21 Series Printers
Self-laminating Label Cartridge for use with BMP21/BMP21-LAB and BMP21-PLUS label printers.
Brand:  Brady™ 110929
Product Code. 16282941
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Abrasion and smudge resistant
  • Moisture resistant and self extinguishing
  • For Communications Cables, Panel Identification, Voice/Data Identification, Wire and Cable Marking
  • Compatible with BBP72 Label Printer, BBP81 Label Printer, Brady 200MVP Plus, Brady 2461, Brady 300MVP Plus, Brady 300X-Plus II, Brady 3481, Brady 360X-Plus II, Brady 600X-Plus II, Brady 6441, BradyPrinter PR Plus Series Label Printer, BradyPrinter PR Plus 600, Tagus T200
Compliance
RoHS Compliant, UL Recognized
Specifications
Specifications
Labeling Ribbon
BMP21-LAB Label Printer, BMP21-PLUS Label Printer, IDPAL Handheld Label Maker, LABPAL Handheld Label Maker, Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer
Thermal Transfer
1.25 in.
31.75 mm
Matte
-70°C to +70°C
Black on White
Self-laminating Vinyl
14 ft.
4.27 m
4.27 m x 31.75 mm (14 ft. x 1.25 in.)
-94°F to +158°F
UL Recognized to UL 2238, RoHS compliant to 2011/65/EU (Pass 2015/863)