Learn More
Brady™ Thermal Transfer Conical Vial Labels
Thermal Transfer Conical Vial Labels are made of self-laminating polyester (B-461), a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
Brand:  Brady™ 062366
 View more versions of this product
Product Code. 11831012
  • 708.00 € / 1 roll
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Self-Laminating Polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • Service temperature of -320 to 230° F (-196 to 110° C)
Specifications
Specifications
RoHS, Pass 2002/95/EC
Acrylic, Permanent
Matte
Polyester
Non-sterile
BBP™81, Brady 1244, Brady 1344, Brady 200MVP Plus, Brady 2461, Brady 300MVP Plus, Brady 300X-Plus II, Brady 3481, Brady 360X-Plus II, Brady 600X-Plus II, Brady 6441, IP300, IP600, Tagus T200, Tagus T300, Thermal transfer printers
3000/Pk.
-196 °C to 110 °C
0.048 mm
1.5 in. H x 3.5 in. W
88.9 mm
1.5 in.
31.75 mm
Thermal Transfer Printable Labels
White, Translucent
Label
Thermal Transfer Printer Label
Bottles, Conical tube, deep well plate, Laboratory identification, Petri dish, Tube, Vial
1
Rectangle
0.0019 in.
41.28 mm
3.5 in.
38.10 mm
38.10 mm