Learn More
Brady™ Thermal Transfer Printable Labels
Nylon cloth labels for thermal transfer printers
Brand:  Brady™ 062365
 View more versions of this product
Product Code. 12069857
  • 296.00 € / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Feature StainerBondz™ super chemical resistant polyester
  • B-481 material, coupled with R6410 series ribbon, will remain resistant when exposed to xylene and other chemicals that are commonly used in the slide staining process
  • No print removal, no label buckling, and no discoloration that would cause bar codes to become unreadable
  • Permanent acrylic adhesive
  • Label stays adhered to the slide
Specifications
Specifications
Thermal Transfer Printable Labels
White
Label
Slide Label
BBP™81, Brady 1244, Brady 1344, Brady 200MVP Plus, Brady 2461, Brady 300MVP Plus, Brady 300X-Plus II, Brady 3481, Brady 360X-Plus II, Brady 600X-Plus II, Brady 6441, IP300, IP600, Tagus T200, Tagus T300, Thermal Transfer Printers
3000/Pk.
-196 °C to 110 °C
19.05 mm
15.24 mm
15.24 mm
Acrylic, Permanent
Matte
Polyester
General Identification, Slide label ID
1
Rectangle
0.048 mm
1.625 in.
0.6 in.
25.40 mm