Learn More
Brady™ Thermal Transfer Printable Nylon Cloth Labels
Thermal Transfer Printable Nylon Cloth Labels with high adhesion for use in autoclave, freezer or liquid nitrogen.
Brand:  Brady™ 019260
306.94 EUR valid until 2025-03-09
Use promo code "24130" to get your promotional price.
 View more versions of this product
Product Code. 10180213
  • 325.00 € / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • White, matte finished labels suitable for use in laboratory identifications and vial labeling.
  • Low profile.
  • Recommended for use with Brady thermal transfer printer ribbon series 4900.
  • Compatible with color Brady series 6200, 6000, 4300, and 4500.
Specifications
Specifications
Autoclavable
THT-163-499-3
Permanent Acrylic, Permanent
Matte
Nylon Cloth
Thermal Transfer
Non-sterile
BBP™81, Brady 1244, Brady 1344, Brady 200MVP Plus, Brady 2461, Brady 300MVP Plus, Brady 300X-Plus II, Brady 3481, Brady 360X-Plus II, Brady 600X-Plus II, Brady 6441, IP300, IP600, Tagus T200, Tagus T300, thermal transfer printers
1
-40 °C
Rectangle with Rounded Corners
1 x 0.375 in.
-40°F to +194°F
80 lb./in.
0.166 mm
1 in.
1 in.
9.53 mm
9.53 mm
UL, CSA C22.2 No.0.15-95, RoHS 2002/95/EC, DIN VDE 0472 part 815
Thermal Transfer Printer Labels
White
Label
Single-side Printable
Labels
For laboratory identification, vial top
Roll of 3000 Labels
2 Years
3000 Pk.
0.375 x 1 x 0.375 in. (L x W x dia.)
2.5 x 0.95 cm
-40°C to +90°C
0.0065 in.
12.70 mm
25.40 mm
0.37 in.
0.375 in.