Learn More
Brady™ and TLS-PC Link™ Tamper-Evident Labels
and TLS-PC Link™ Labels are made of Tamper-Resistant Vinyl (B-351) that is designed to fracture easily in order to show signs of product tampering and to prevent on-piece label removal.
Brand:  Brady™ 018700
Product Code. 10542402
Description
Description
  • High performance white matte vinyl with permanent adhesive is ideal for applications that require resistance to product tampering
  • Use caution when removing from liner as material is fragile
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • Service temperature of -40 to 180° F (-40 to 82° C)
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ and TLS-PC Link™ Tamper-Evident Labels > 2 x 1 in.