Learn More
Brady™ ToughStripe™ Floor Marking Center Marks
ToughStripe™ Floor Marking Center Marks are extremely durable die-cut floor marking shapes made of rugged polyester with ultra-aggressive adhesive and hold up against forklift traffic.
Brand:  Brady™ 104469
 View more versions of this product
Product Code. 11711246
  • 40.55 € / 1 pack
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Visible floor-marking improves safety and organization in almost any industrial environment
  • High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
  • Ideal for marking corners of work cells and storage locations
  • Can be easily applied by one person working alone
Specifications
Specifications
ASTM ANSI F1679, ANSI A1264.2 2006, OSHA 1910.22
ToughStripe Diecut Shapes (L's, T's, +'s, footprints and more)
Yellow
Polyester
For floor corner mark, Used to mark aisles, passageways, storage locations and point out safety equipment and exits
20/Pk.
0°F to 130°F
0.008 in.
5 in.
2 in.
127.00 mm
TS-50.80-514-CEN-YL-20
Rubber, Permanent
Tape
Non-sterile
115 Gardner Units
Cross
-18°C to +54°C
0.201 mm
12.7 cm
50.8 mm