Learn More
Brady™ ToughStripe™ Floor Marking Tape
ToughStripe™ Floor Marking Tape (3.5m/100 ft. in length) is a long-lasting, heavy-duty solution for marking floors, aisles, work cells and other designated areas.
Brand:  Brady™ 104379
 View more versions of this product
Product Code. 11781695
  • 155.00 € / 1 roll
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Visible floor-marking improves safety and organization in almost any industrial environment
  • High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Can be easily applied by one person working alone
Specifications
Specifications
ASTM ANSI F1679, ANSI A1264.2 2006, OSHA 1910.22
ToughStripe FLOOR TAPES
Black, White
Tape
Non-sterile
115 Gardner Units
0°F to 130°F
0.008 in.
100 ft. x 4 in. (30.48 m x 10.2 cm)
30.48 m
10.2 cm
TS-101.60-514-BK/WT-STR-RL
Rubber, Permanent
Gloss
Polyester
Used to mark aisles, passageways, storage locations and point out safety equipment and exits
1 Roll
-18°C to +54°C
0.201 mm
100 ft.
4 in.