Learn More
Brady™ ToughStripe™ Labels with Overlaminate
Create custom-printed floor labels quickly and easily
Brand:  Brady™ 142154
 View more versions of this product
Product Code. 11711006
  • 882.00 € / 1 roll
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Includes:
  • 1 roll of B-483 polyester
  • 1 roll of B-634 polyester overlaminate
Description
Description
Easy Three-Step Process
  • Print label
  • Position on floor
  • Apply overlaminate
Specifications
Specifications
RoHS 2002/95/EC
GlobalMark ToughStripe Floor Marking Tape with Overlaminate
White, Clear
Tape
For high adhesion to textured metals and low surface energy Plastics
1 Roll of tape and 1 roll of over laminate
0°F to 130°F
0.004 in.
100 ft. x 2.25 in. (30.48 m x 5.71 cm)
30.48 m
5.71 cm
GMK B-483 WT 57,2mm & B-634OLam
Rubber, Permanent
Gloss
Polyester
GlobalMark™ 2 and BMP™ 71 label printer
1 Kit
-18°C to +54°C
0.203 mm
100 ft.
2.25 in.