Learn More
Brady™ Vial Top Labels
Durable Freezerbondz™ Polyester (B-492) Vial Top Labels are designed for laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Brand:  Brady™ 142919
 View more versions of this product
Product Code. 13573310
  • 149.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • B-490 labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • Matte white polyester material has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance of -320 to 212° F (-196 to 100° C)
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Specifications
Specifications
B33-144-492
Acrylic, Permanent
Matte
Laboratory Label
Use in laboratory identification such as vials, centrifuge tubes, test tubes, straws and slides
15.24 mm
1500/Pk.
-196°C to +130 °C
0.081 mm
0.5 x 0.5 in.
12.70 mm
12.70 mm
1.27 cm
FreezerBondz Labels for the BBP33 Printer
White
Polyester
Non-sterile
BBP™33
3
LabelMark, MarkWare
0.0032 in.
15.88 mm
0.5 in.
0.5 in.
0.5 in.