Learn More
Brady™ BradyJet Laboratory Vinyl Labels
Indoor-Use General Purpose Vinyl labels with specially-formulated topcoat for improved durability and color
Brand:  Brady™ 150546
Product Code. 17180391
  • 401.00 € / Pack of 1000
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • B-2595 Indoor-Use General Purpose Vinyl from Brady features a specially-formulated topcoat for improved durability and color
  • Specially-formulated topcoat enhances colors and label appearance
  • Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
  • Topcoat protects print from smudging and harsh conditions
  • Inkjet-receptive labels for use with the BradyJet™ J2000 Color Label Printer
  • 1000 per roll
  • Specifications
    Specifications
    Acrylic
    Matte
    BradyJet™ J2000
    1000/Pk.
    0.0054 in.
    1 in.
    White
    Vinyl
    Secondary Label is Not Included
    1 x 3 in.
    3.2 in.