Learn More
Brady™ BradyJet™ Pre-Cut Vinyl Labels
Brady™ BradyJet™ Pre-Cut Vinyl Labels are ideal for a wide range of identification applications.
Brand:  Brady™ 150032
 View more versions of this product
Product Code. 16150491
  • 485.00 € / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Constructed of indoor-use general purpose vinyl
  • Features a specially-formulated topcoat for improved durability and color
  • Specially-formulated topcoat enhances colors and label appearance
  • Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
  • Topcoat protects print from smudging and harsh conditions
  • Inkjet-receptive labels for use with the BradyJet™ J2000 Color Label Printer
Specifications
Specifications
Acrylic
Matte
Vinyl
BradyJet™ J2000 Printer
Rectangle
-40°C to +60°C
4.13 in.
1.88 in. I.D.
White
Roll
Pipe and Valve Marking; Safety and Compliance ID; Visual Workplace Labeling; Warehouse Marking; Lockout Tagout Labeling; GHS and Chemical Labels; Arc Flash Labels
200/Roll
−40 to 140°F
6.10 x 4.13 in.
6.10 in.