Learn More
Thermo Scientific Chemicals Bradykinin
Brand: Thermo Scientific Chemicals J63131.MA
71.30 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Bradykinin is used to lower blood pressure, increase bradykinin (by inhibiting its degradation) further lowering blood pressure. Bradykinin dilates blood vessels via the release of prostacyclin, nitric oxide, and endothelium-derived hyperpolarizing factor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
HPLC | |
Powder | |
1060.23 | |
10 mg | |
Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg-OH |
BK | |
An endogenous vasodilator peptide | |
Bradykinin | |
RUO - research use only |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y03J029 | Certificate of Analysis | 15/11/2023 | J63131.MA |
U30I003 | Certificate of Analysis | 27/11/2022 | J63131.MA |
Z17H039 | Certificate of Analysis | 12/04/2022 | J63131.MA |
S26B011 | Certificate of Analysis | 08/12/2021 | J63131.MA |
L13Z050 | Certificate of Analysis | 08/12/2021 | J63131.MA |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only