missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ BradyPrinter i7100 - UK with Peel function and vial label applicator
Industrial thermal transfer benchtop printer is ideal for customers requiring reliable high volume printing.
Brand: Brady™ 307654
6925.50 EUR valid until 2025-12-31
Use promo code "24308" to get your promotional price.
This item is not returnable.
View return policy
Description
- The BradyPrinter i7100 industrial thermal transfer benchtop printer is ideal for customers requiring reliable high volume printing.
- Automate vial tube identification with the Vial Label Applicator that prints and applies labels on vial tubes in less than 2 seconds.
- This printer comes with a free Brady WorkStation Basic Design Software Suite, enabling you to immediately start designing professional labels.
Specifications
Color Touchscreen | |
252.00 mm | |
520.00 mm | |
Monochrome | |
150.00 mm/s | |
AC Adaptor |
Thermal Transfer | |
288.00 mm | |
520.00 x 252.00 x 288.00 mm | |
Peel and Present module pre-installed. It ships with a vial label applicator, a UK power cord, a Brady Workstation Basic Suite, 2 empty ribbon cores, a USB cable, a product CD holding the driver, the user manual in languages, the configuration/settings manual in English and a printed user manual in English | |
300 dpi | |
I7100-300-P-UK-VLA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction