Learn More
STEMCELL Technologies BrainPhys™ Neuronal Medium and SM1 Kit
Kit including BrainPhys™ Neuronal Medium and SM1 Neuronal Supplement for culture of primary and ES/iPS cell-derived neurons
Brand:  STEMCELL Technologies 05792
Encompass
Product Code. 17108301
  • 291.00 € / 500mL
Explore more special offers
Description
Description
  • Promote, rather than inhibit, neuronal activity and maturity in your cultured primary or human pluripotent stem cell (hPSC)-derived neurons
  • Based on the formulation by Bardy and Gage (Bardy et al. PNAS, 2015), BrainPhys serum-free neuronal basal medium is optimized to yield a higher proportion of synaptically active neurons by mimicking the central nervous system (CNS) extracellular environment
  • Use BrainPhys Neuronal Medium for long-term culture of hPSC- and CNS-derived neurons and to perform functional assays, such as microelectrode array-based recordings or live-fluorescent imaging, without shocking your cells by changing media
  • To ensure cell survival in long-term culture, BrainPhys must be combined with an appropriate supplement
  • For your convenience, BrainPhys Neuronal Medium N2-A & SM1 Kit is available for hPSC-derived neurons and can be combined with lineage-specific growth factors and/or small molecules
Specifications
Specifications
Neural Cells, PSC-Derived, Neurons, Pluripotent Stem Cells
Cell Culture; Differentiation; Maintenance
500 mL Kit