Learn More
STEMCELL Technologies BrainPhys™ Without Phenol Red
Serum-free neurophysiological basal medium for improved neuronal function
Brand:  STEMCELL Technologies 05791
Additional Details : Weight : 0.50000kg
Product Code. 17198291
  • 189.00 € / 500mL
Estimated Shipment: 09-07-2024
to see stock.
Description
Description
  • Promote, rather than inhibit, neuronal activity and maturity in your cultured primary or human pluripotent stem cell (hPSC)-derived neurons in a phenol red-free environment
  • Based on the formulation by Bardy and Gage (Bardy et al. PNAS, 2015), BrainPhys serum-free neuronal basal medium is optimized to yield a higher proportion of synaptically active neurons by mimicking the central nervous system extracellular environment
  • Use BrainPhys Without Phenol Red to culture primary neurons, differentiate and mature hPSC-derived neurons, record microelectrode array-based neuronal activity, live-fluorescent image neurons, and transdifferentiate somatic cells to neurons without hormonal signaling
  • To ensure cell survival in long-term serum-free culture, BrainPhys Without Phenol Red must be combined with an appropriate serum-replacement supplement, such as NeuroCult SM1 Neuronal Supplement (Catalog #05711) and/or N2 Supplement-A (Catalog #07152)
Specifications
Specifications
Neural Cells, PSC-Derived, Neurons, Pluripotent Stem Cells
Cell Culture; Differentiation; Maintenance
500 mL