missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Four-Clear Sided Disposable Polystyrene Cuvettes for Fluorescence
German manufactured premium cuvettes
Brand: BRAND™ 759035
93.25 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- BRAND premium quality four sided clear disposable cuvettes for best results in fluorescence applications.
- Choose polystyrene for general purpose requirements in the visible range (340-800nm).
- Choose proprietary UV-plastic cuvettes with virtually no autoflourescence for best signal to noise ratio for assays in the 230-800nm range.
- 10mm lightpath to fit most fluorimeters.

Specifications
Fluorimeter Cuvette | |
2.5 to 4.5 mL | |
12.5mm | |
10 mm | |
fluorometers | |
Fluorimetry |
BRAND four sided clear cuvettes for fluorimetry, polystyrene, pack of 500 | |
Polystyrene | |
500 | |
45 mm | |
12.5 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction