missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ 96-well Deep-well Plates
These deep-well plates are stackable for easy storage.
Brand: BRAND™ 701352
109.52 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
This item is not returnable.
View return policy
Description
- DNA-, DNase- and RNase-free
- Designed for a wide range of applications, such as High-Throughput Screening (HTS), tests requiring a mother plate, cell and tissue cultures, serial dilutions, reagent transfer and sample storage down to -80°C
- Deep-well plates are manufactured according to ANSI/SLAS 1 and 4 in.
- Choose PP plates for autoclavability and chemical resistance.
- Choose PS plates for transparency and surface properties.
- Deep Well Plates are ideal for sample storage, HTS assays requiring a motherplate, cell and tissue culture, immunological assays, and other applications.

Specifications
Standard | |
Not Autoclavable | |
Polystyrene | |
96 | |
Flexible mat covers or adhesive sealing films to reduce sample evaporation and contamination | |
U-bottom |
8 x 12 | |
Deep-well Plate | |
1.1 mL | |
Round Bottom (U-well) | |
32 | |
32 Pk. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ 96-well Deep-well Plates > Well Volume: 1.1 mL, Material: Polystyrene, Bottom: Round
Spot an opportunity for improvement?Share a Content Correction