missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ 96-well Deep-well Plates

These deep-well plates are stackable for easy storage.

Brand:  BRAND™ 701342

134.16 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.


 View more versions of this product

Product Code. 15613987

  • 166.00 € / 1 pack
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • DNA-, DNase- and RNase-free
  • Designed for a wide range of applications, such as High-Throughput Screening (HTS), tests requiring a mother plate, cell and tissue cultures, serial dilutions, reagent transfer and sample storage down to -80°C
  • Deep-well plates are manufactured according to ANSI/SLAS 1 and 4 in.
  • Choose PP plates for autoclavability and chemical resistance.
  • Choose PS plates for transparency and surface properties.
  • Deep Well Plates are ideal for sample storage, HTS assays requiring a motherplate, cell and tissue culture, immunological assays, and other applications.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
Autoclavable
Polypropylene
96
Round Bottom (U-well)
Elevated Rim
8 x 12
Deep-well Plate
1.2 mL
Non-sterile
U-bottom
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5057417-10 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog Number701342
Lot Number5057417 Certificate TypeCertificate of Analysis Date20/11/2023 Catalog Number701342
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ 96-well Deep-well Plates > Well Volume: 1.2 mL, Material: Polypropylene, Bottom: Round

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.