missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Self-adhesive Plate Sealing Film
Securely seals multiwell plates. Self-adhesive Plate Sealing Film can be applied directly to plates and removed without the use of expensive equipment, or can be applied with a hard rubber roller to ensure secure and uniform attachment of the film to the plate.
60.70 € - 205.00 €
Product Code | Brand | Material | For Use With (Application) | No. per Pack | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | For Use With (Application) | No. per Pack | Price | Quantity & Availability | |||
10655861
|
BRAND™
781390 |
Polyester | ELISA, PCR | 100 Sheets |
146.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
10135623
|
BRAND™
701370 |
Polyethylene, Polypropylene | Automation | 50 Sheets |
192.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||
10185763
|
BRAND™
701371 |
Vinyl | Storage, fluorescence measurements | 50 Sheets |
68.60 €
Pack of 50 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||
10145813
|
BRAND™
701372 |
Vinyl | Luminescence measurements | 50 Sheets |
60.70 €
Pack of 50 |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
11673119
|
BRAND™
701367 |
Polypropylene | Storage | 100 Sheets |
123.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
15660499
|
BRAND™
701374 |
Vinyl | Automation | 100 |
159.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
10135371
|
BRAND™
701364 |
Rayon | Cell and tissue culture | 100 Sheets |
125.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
10767564
|
BRAND™
781391 |
Polyester | ELISA, Real-time PCR | 100 Sheets |
205.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
10182962
|
BRAND™
781381 |
Aluminum | Cold storage | 100 Sheets |
150.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
10599892
|
BRAND™
701365 |
Rayon | Cell and tissue culture | 50 Sheets |
113.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||
Description
- Easily apply to plates and remove without the use of expensive equipment.
- Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
- Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
- Polyethylene
- Polyester
- Aluminum
- Rayon
- Polypropylene
- Vinyl
Specifications
Sealing Film |