missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ seripettor™ pro Bottletop Dispensers

Designed to handle a wider range of liquids than the regular seripettor

Brand:  BRAND™ 4720420

 View more versions of this product

Product Code. 10419894

  • 384.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket

Includes: Each seripettor pro bottletop dispenser is supplied with discharge tube, filling tube, spare dispensing catridge, mounting tool, two polypropylene adapters (GL45/33 and GL45/38), operating manual, and one-year warranty.
This item is not returnable. View return policy

Description

Description

Easily-replaceable wiping seal dispensing cartridge is especially suitable for liquids that tend to form crystals

  • Quick-change cartridge makes service a breeze and keeps life-time service costs low
  • Pump assembly with corrosion resistant spring has opaque upper sleeve that protects liquid from UV-light during dispensing operations
  • Telescoping filling tube adjusts to fit many bottles without cutting
  • Threaded safety cap attaches and removes with a quick twist
  • Spring-loaded piston permits one-handed, semi-automatic operation

Liquids unsuitable for the seripettor pro include, but are not limited to:

  • Hydrofluoric acid, or other chemicals that attack high-density polyethylene,PTFE, ETFE, FEP, PFA, alumina-ceramic, platinum-iridium, and borosilicate glass
  • Carbon disulfide
  • Density exceeding 2.2g/cm
  • Viscosity exceeding 300mm2/sec (2mL instrument), 150mm2/s (10mL instrument), 75mm2/s (25mL instrument)
  • Vapor pressure up to 500mbar
Temperature of instrument and reagents must be between 59° and 104°F (15° and 40°C).
Agar culture media to 140°F (60°C).

Order Info

Replaceable piston and cylinder are permanently connected as a cartridge module for safety. If piston, cylinder, or seal are damaged, simply replace cartridge in seconds without tools.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

≤±1.2%
Up to 2.2 g/cm3
Discharge tube, telescoping filling tube, spare dispensing cartridge, mounting tool, GL45/33mm and GL45/38mm polypropylene adapters, operating manual
≤0.2%
15°C to 40°C
1000mm2/sec.
Seripettor Pro Bottletop Dispenser
0.2 to 2 mL
0.04 mL
59°F to 104°F
Up to 500mbar
1 Year
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.