missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Skirted Low-Profile PCR Plates
Manufactured from high-quality virgin plastics without lubricants or chemical additives that can contaminate reagents and samples. Provide excellent, reproducible results in most PCR applications.
Brand: BRAND™ 781377
262.85 EUR valid until 2025-12-31
Use promo code "24286" to get your promotional price.
This item is not returnable.
View return policy
Description
- Autoclavable
- Free of DNA, DNase, RNase, and other PCR inhibitors.
- Thin, uniform walls ensure consistent, precise thermal transfer and excellent results.
- Low profile, skirted plates from BRAND are compatible with many major brands of thermal cyclers, including BIORAD™ CFX 96, Agilent AriaMx, Amersham, Beckman™ CEQ, PEQLAB™, Techne™ TC-412, PCRmax, Flexi, Eppendorf™ Mastercycler™ and other systems
- White PCR plates are uniformly colored with titanium dioxide (TiO2) and have smooth surfaces to optimize reflection of the fluorescence signal.

Specifications
Standard | |
Autoclavable | |
96 | |
qPCR | |
Low |
8 x 12 | |
Clear | |
Yes | |
50 | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ Skirted Low-Profile PCR Plates > Clear; Volume: 0.15mL
Spot an opportunity for improvement?Share a Content Correction