Learn More
BRAND™ White 96-well PCR Plates
GREENER_CHOICE
Designed specifically to optimize results when performing quantitative Real Time PCR (qPCR)
Brand:  BRAND™ 781354
218.25 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10034564
  • 239.00 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
White PCR strips and plates offer significantly better results in the analysis of fluorescence signals and consequently with the quantification of replicated DNA
  • DNase-, DNA- and RNase-free
  • Smooth surfaces to optimize relection of the fluorescence signal
  • Suitable for use in common thermal cyclers
  • Extra-thin wall dimensions to provide the optimal thermal transfer and short cycle times
  • Manufactured under the most modern clean room conditions
  • Uniformally colored with titanium dioxide (TiO2)
  • Blue embossed alphanumeric coding and cut corner design to to facilitate orientation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
8 x 12
Polystyrene
Non-sterile
Optimize results when performing quantitative RT-PCR
Without Lid
White
96
0.2 mL
No Skirt