Learn More
BRAND™ White Real-Time PCR (qPCR) 96-Well Microplates
Designed specifically to optimize results when performing quantitative Real Time PCR (qPCR)
Brand:  BRAND™ 781365
298.32 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.
 View more versions of this product
Product Code. 11658439
  • 388.00 € / Pack of 50
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
White PCR strips and plates offer significantly better results in the analysis of fluorescence signals and consequently, the quantification of replicated DNA.
  • DNase-, DNA- and RNase-free
  • Smooth surfaces to optimize reflection of the fluorescence signal
  • Suitable for use in common thermal cyclers
  • Extra-thin wall dimensions to provide the optimal thermal transfer and short cycle times
  • Manufactured under the most modern cleanroom conditions
  • Uniformly colored with titanium dioxide (TiO2)
  • White; Low-profile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNA, DNase, RNase-free
8 x 12
With sealing film
96
50/Pk.
Standard
White
Polypropylene
PCR