missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates™ pureGrade™ 384-Well Microplates

Non-treated microplates for diverse applications

Brand:  BRAND™ 781622

158.31 EUR valid until 2024-12-31
Use promo code "21894" to get your promotional price.


 View more versions of this product

Product Code. 10236333

  • 203.00 € / Pack of 50
Estimated Shipment: 26-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Non-treated PS surface
  • Medium binding, non-sterile
  • Standard plate for most applications
  • Free from endotoxins, DNase, DNA, RNase; noncytoxic

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
16 x 24
Polystyrene
384
Non-treated
For homogenous assays, screenings, storage
Without Lid
Black
0.1 mL
Non-sterile
Flat Bottom (F-well)
50/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers