Learn More
BRAND™ BRANDplates™ pureGrade™ 96-Well Microplates
Constructed of polystyrene with nontreated surface. BRANDplates™ pureGrade™ 96-Well Microplates are noncytotoxic, and free from endotoxins, DNase, DNA, and RNase. Medium binding, nonsterile microplates are suited for homogenous assays, screening, and storage with gray alphanumeric embossing on white and black solid plates for easy well identification.
Brand:  BRAND™ 781605
274.60 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10732694
  • 314.00 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polystyrene surface
  • Medium binding, nonsterile
  • Standard plate for most applications
  • Gray alphanumeric embossing on white and black solid plates for easy well identification
  • Strip plates available
  • Standard plates available in U-, V-, F-, C-shapes
  • Transparent bottoms available in F- shape
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase; noncytotoxic
  • Suited for homogenous assays, screening, and storage.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Includes Barcode
Standard
8 x 12
White
350 μL
Non-sterile
Flat Bottom (F-well)
Homogenous assays, screenings and storage
Yes
ANSI, SLAS
Without Lid
Medium Binding
Polystyrene
96
Non-treated
Yes
100/Cs.