missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates™ pureGrade™ 96-Well Microplates
Constructed of polystyrene with nontreated surface. BRANDplates™ pureGrade™ 96-Well Microplates are noncytotoxic, and free from endotoxins, DNase, DNA, and RNase. Medium binding, nonsterile microplates are suited for homogenous assays, screening, and storage with gray alphanumeric embossing on white and black solid plates for easy well identification.
Brand: BRAND™ 781615
698.06 EUR valid until 2024-12-31
Use promo code "21894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Polystyrene surface
- Medium binding, nonsterile
- Standard plate for most applications
- Gray alphanumeric embossing on white and black solid plates for easy well identification
- Strip plates available
- Standard plates available in U-, V-, F-, C-shapes
- Transparent bottoms available in F- shape
- Available in clear, white or black
- Free from endotoxins, DNase, DNA, RNase; noncytotoxic
- Suited for homogenous assays, screening, and storage.
Specifications
Includes Barcode | |
Standard | |
Medium Binding | |
UV Transparent | |
350 μL | |
Non-sterile | |
Flat Bottom (F-well) | |
Homogenous assays, screenings and storage | |
Yes |
ANSI, SLAS | |
8 x 12 | |
Black | |
Polystyrene | |
96 | |
pureGrade | |
Yes | |
50/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction