Learn More
BRAND™ Polypropylene Thread Adapters for Bottle-Top Dispensers
Adapt current model Dispensette™ seripettor™, Titrette™ and QuikSip™ to common bottle sizes.
Brand:  BRAND™ 704397
14.23 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
 View more versions of this product
Product Code. 10566522
  • 15.20 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polypropylene Construction
  • For use with Dispensette™ seripettor™ Titrette™ and QuikSip™ Bottle-top dispensers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bottle Thread Adapter
Polypropylene
Dispensette™, seripettor™, Titrette™ and QuikSip™
GL 45/38