Learn More
BRAND™ Polypropylene Thread Adapters for Bottle-Top Dispensers
Adapt current model Dispensette™ seripettor™, Titrette™ and QuikSip™ to common bottle sizes.
Brand:  BRAND™ 704328
 View more versions of this product
Product Code. 10568911
  • 9.65 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polypropylene Construction
  • For use with Dispensette™ seripettor™ Titrette™ and QuikSip™ Bottle-top dispensers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bottle Thread Adapter
Polypropylene
Dispensette™, seripettor™, Titrette™ and QuikSip™
GL 33/28