missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Centrifuge Tube Mini-Cooler

Product Code. 10441063 Shop All Brand Products
Click to view available options
Color:
Red
White
Yellow
Holds Temp., Hours:
-20°C for 60 min.
-70°C for 45 min.
0°C for 60 min.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10441063

Brand: BRAND™ 114940

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench.

  • Manufactured from durable polycarbonate
  • Filled with non-toxic gel
  • Hold twelve 0.5 to 2.0 mL tubes
  • Provided individually
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Centrifuge tube
Holds Temp., Hours -70°C for 45 min.
Material Polycarbonate
Type Mini Cooler
Color White
Capacity (Metric) 0.5 to 2 mL
Holds 0.5 to 2 mL Tubes
Autoclavable Not Autoclavable
Temperature Control Passive
Temperature Range -70°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.