missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Transferpette™ Single Channel Electronic Pipets
GREENER_CHOICE

Product Code. 10485802 Shop All Brand Products
Click to view available options
Volume (Metric):
0.5 to 10 μL
10 to 200 μL
100 to 1000 μL
2 to 20 μL
250 to 5000 μL
50 to 1000 μL
500 to 5000 μL
Includes:
EU plug
Europe AC Adapter
UK/Ireland AC Adapter
UK/Ireland plug
Without AC Adapter
Unit Size:
Each
Color:
Blue
Gray
None
Red
Yellow
This item is not returnable. View return policy

Product Code. 10485802

Brand: BRAND™ 705307

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Combines comfortable design, balanced weight distribution, intuitive software and user-friendly documentation for your pipetting needs.

  • Functional, ergonomic housing design
  • Individually adjustable finger rest
  • Intuitive menu structure and comprehensively illustrated user manual
  • Significantly reduced tip attachment and ejection forces using universal tip
  • Corrosion-resistant piston and ejector
  • Five convenient programs: pipetting, reverse pipetting, mixing, GEL-Electrophoresis, dispensing
  • Ready for use: 4000 pipetting cycles with each battery charge, battery refresh function even during recharging
  • Each pipette has a DE-M marking and is supplied with performance certificate
  • Supplied with battery and silicone oil
TRUSTED_SUSTAINABILITY

Specifikationer

Accuracy ±30μL
Autoclavable Autoclavable
Number of Channels 1
Product Line Transferpette
Type Electronic Pipette
Volume (Metric) 250 to 5000 μL
Includes EU plug
Plug Type EU
Temperature (Metric) Operating 15°C to 40°C
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.