missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ seripettor™ Bottletop Dispensers

Suitable for use with bases, buffers, aqueous solutions of organic and inorganic substances, dilute non-oxidizing acids, and many polar solvents. Space-saving, low-rise design provides stability for easy one-handed operation and fit most common reagent bottles with the included adapters.

215.00 € - 255.00 €

Specifications

Product Type Seripettor Bottletop Dispenser
Accuracy ≤±1.2%
Density Up to 2.2 g/cm3
Includes Discharge tube, telescoping filling tube, spare dispensing cartridge, 45/33mm and 45/38mm thread polypropylene adapters, operating manual
Max. Pressure 500 mbar
View More Specs

Includes: Two dispensing cartridges, discharge tube, filling tube, two polypropylene thread adapters (45/33 and 45/38) and operating manual

Products 3
Product Code Brand Volume (Metric) Increments Viscosity Rating Price Quantity & Availability  
Product Code Brand Volume (Metric) Increments Viscosity Rating Price Quantity & Availability  
10309744
View Documents
BRAND™
4720120
0.2 to 2 mL 0.04 mL 1000mm2/sec.
215.00 €
Each
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
 
10320861
View Documents
BRAND™
4720140
1 to 10 mL 0.2 mL 150mm2/sec.
215.00 €
Each
In Stock
to see stock.
Add to basket
 
10598882
View Documents
BRAND™
4720150
2.5 to 25 mL 0.5 mL 75 mm2/sec.
255.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Suitable for use with bases, such as ammonia and sodium hydroxide; buffers; aqueous solutions of organic and inorganic substances; dilute non-oxidizing acids; and many polar solvents.

  • Fluid path consists of materials such as polyethylene, polypropylene and EPDM (ethylene-propylene-diene-rubber)
  • Full scale accuracy of ≤±1.2% and reproducibility of ≤0.2%
  • Fit most common reagent bottles with the included adapters
  • Space-saving, low-rise design provides stability for easy one-handed operation
  • With simple, clear volume adjustment for accuracy and a sturdy, supported discharge tube for safe operation
  • No fragile glass parts to break
  • Replaceable cylinder/piston cartridge has a wiping seal for renewability

This dispenser is not intended for use with scintillation fluid; non-polar solvents, such as xylene and most xylene substitutes; or strong, oxidizing or concentrated acids. For these liquids, choose the appropriate Dispensette™ bottletop dispenser.

Order Info

Unit disassembles in seconds for easy cleaning and maintenance. Replaceable piston and cylinder are permanently connected as a cartridge module for safety. If piston, cylinder, or seal are damaged, simply replace cartridge in seconds without tools.

Specifications

Specifications

Seripettor Bottletop Dispenser
Up to 2.2 g/cm3
500 mbar
59°F to 104°F
1 Year
≤±1.2%
Discharge tube, telescoping filling tube, spare dispensing cartridge, 45/33mm and 45/38mm thread polypropylene adapters, operating manual
≤0.2%
15°C to 40°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.