missing translation for 'onlineSavingsMsg'
Learn More

Branson Ultrasonics™ Sonifier™ Continuous Flow Attachment for Models 250, 350, and 450

Processes low-viscosity solutions at rates up to 30lph

Brand:  Branson Ultrasonics™ 100-146-171

Product Code. 10467813

  • 2500.00 € / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Can be used to emulsify, disperse, and homog-enize by pumping a solution through a zone of intense ultrasonic activity.

  • Degree of processing is controlled by varying the amplitude of the ultra-sonic horn and the flow rate
  • Some solutions may require recirculating until the desired results are obtained
  • Incorporates a cooling jacket through whicha suitable cooling liquid can be circulated to retard heat build-up during extended operation
  • May also be sealed in a closed system to assure sterile conditions and inhibit contamination when working with infectious materials
  • Overall dimension (L x Dia.): 12.7 x 9.65cm (5 x 3.8 in.)

  • Production of vaccines and antigens
  • Emulsification of immiscible liquids (with or w/o surfactants)
  • Removal of cell walls
  • Dispersing metal oxides in solvents

Specifications

Specifications

Continuous Flow Attachment
9.65 cm
Emulsify, disperse and homogenize by pumping a solution through a zone of intense ultrasonic activity
5 in.
Stainless Steel
2.26 kg
3.8 in.
Ultrasonic Cell Disruptor
Cooling jacket
12.7 cm
5 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.