Learn More
BD Biosciences™ Brefeldin A Solution (RUO/GMP)
Inhibits the secretion of newly synthesized cytokine and CD69.
Brand:  BD Biosciences™ 347688
Additional Details : Weight : 0.05300kg
Product Code. 15806118
  • 348.00 € / Each
Estimated Shipment: 23-07-2024
to see stock.
Description
Description
  • Inhibits the secretion of newly synthesized cytokine and CD69.
  • Includedcomponent of BD FastImmune CD4 Intracellular Cytokine Detection Kit
Specifications
Specifications
250 μL