Learn More
Hirschmann™ Bright Lined Counting Chambers
Includes 2 haemocytometer glass covers.
Brand:  Hirschmann™ 8100203
 View more versions of this product
Product Code. 10469132
  • 170.00 € / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Double net ruling
  • Bright lined
  • With LOT number on packaging
  • Labelled with: usable until, CE pursuant to IVD 98/79 EC
Specifications
Specifications
Counting Chamber (Neubauer)
0.1 mm
CE pursuant to IVD 98/79 EC
2 haemocytometer glass covers