Learn More
Hirschmann™ Bright Lined Counting Chambers
Click to view available options
Product Type:
Counting Chamber (Burker)
Counting Chamber (Fuch-Rosenthal)
Counting Chamber (Neubauer)
Counting Chamber (Thoma)
Unit Size:
Each
Description
- Double net ruling
- Bright lined
- With LOT number on packaging
- Labelled with: usable until, CE pursuant to IVD 98/79 EC
Specifications
Specifications
| Product Type | Counting Chamber (Thoma) |
| Depth (Metric) | 0.1 mm |
| Certifications/Compliance | CE pursuant to IVD 98/79 EC |
| Includes | 2 haemocytometer glass covers |