Learn More
Invitrogen™ BrightComp eBeads™ Compensation Bead Kits
BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.
Brand:  Invitrogen™ A54742
 View more versions of this product
Product Code. 17817071
  • 262.00 € / Pack of 25
Explore more special offers
Description
Description
BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.
• Three levels of intensity, perfect for any samples no matter the level of expression
• Near-identical, spectrally-matched dye to fluorescent protein
• Consistent, reliable performance
Accurate compensation is crucial for effective multicolor analysis in flow cytometry. Each kit provides polystyrene microspheres with near-identical, spectrally-matched fluorescent protein dye as well as negative control beads in a single dropper bottle.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CFP BrightComp eBeads™ Compensation Bead Kit
25 tests
Compensation Beads
405 nm
Flow Cytometry