Learn More
Brij™ 35, 30 w/v % solution in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 329581000
24.98 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 29
Additional Details : CAS Number : 9002-92-0 Weight : 0.10300kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Brij™ 35 | |
9002-92-0,7732-18-5 | |
1.0300g/mL | |
C12H25(OCH2CH2)23OH | |
100 mL | |
15, 7675 | |
1199.56 | |
Glass bottle |
Colorless | |
Jelly Substance at 25°C | |
C58H118O24 | |
MFCD00080891 | |
06,70 | |
Solubility in water: soluble. | |
29 to 31% | |
1.03 |
RUO – Research Use Only