missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brij™-35, 30% Solution

Stabilized Brij-35 detergent provided as 10% and 30% solutions for use in various protein methods including cell lysis and HPLC.

Brand:  Thermo Scientific™ 20150


Product Code. 10538893

  • 216.00 € / 950mL
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

This Thermo Scientific formulation of Brij 35 is a stabilized 30% (w/w) solution of Brij-35 detergent for use in various protein methods. Brij 35 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or a surfactant in various HPLC applications.

Properties of Brij 35 Detergent:

• Molecular Weight: 1225 g
• Detergent Class: Non-ionic
• Aggregation Number: 40
• Micelle Molecular Weight: 49,000 g
• Critical Micelle Concentration (CMC): 0.09 mM (0.011%, w/v)
• Cloud Point: >100°C
• Dialyzable: No

Features of this Thermo Scientific detergent solution:

• A nonionic detergent for use in various protein methods
Accurate—30% detergent solution in ultrapure water
Easy-to-use—solutions are simple to dispense and dilute for use

Specifications of this Thermo Scientific Brij 35 30% solution:

• Visual: Clear, colorless viscous liquid, free of foreign material.
• Concentration: 27.0 to 30.0%

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Brij-35, 30% Solution
Store at Room Temperature
Brij™, Surfact-Amps™
950 mL
Detergent
Liquid
Detergent Solution
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Brij-35, 30% Solution

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Dodecan-1-ol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.