Learn More
Proteintech Broad Range Prestained Protein Marker
The PL00002 Broad Range Protein Marker is a ready-to-use three-color protein standard with 13 prestained proteins covering a wide range of molecular weights from 3 to 245 kDa in Tris-Glycine buffer.
Brand:  Proteintech PL00002-100UL
Product Code. 17308262
  • 69.00 € / 100µL
Explore more special offers
Description
Description
  • The PL00002 Broad Range Prestained Protein Marker is designed for monitoring protein separation during SDS-polyacrylamide gel electrophoresis, verification of Western transfer efficiency on membranes (nitrocellulose, PVDF, or nylon) and for estimating the size of proteins.
  • Contents: Approximately 0.1~0.4 mg/mL of each protein in the buffer (20 mM Tris-phosphate (pH 7.5 at 25°C), 2 % SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, and 15 % (v/v) Glycerol).
Specifications
Specifications
Colorimetric
3 to 245
100 Tests
13
Blue, Red, Green
Aliquot and store at -20™C for up to 1 year.
Product Suggestions
Product Suggestions
Videos
SDS Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Proteintech Broad Range Prestained Protein Marker > 100 Tests