missing translation for 'onlineSavingsMsg'
Learn More
Bromate/Bromide Solution, Fisher Chemical™ Product Code.: 12980914

Bromate/Bromide Solution, Fisher Chemical™

Product Code. 12980914
1000 mL
Click to view available options
Quantity:
1000 mL
This item is not returnable. View return policy

Product Code. 12980914

Brand: Fisher Chemical VL 211 1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Quantity 1000 mL
Concentration or Composition (by Analyte or Components) 0.05mol/L
compliance-icons
Product Identifier
  • Bromine Solution - Bromate/Bromide 0.05M (0.1N) Volumetric solution
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : potassium bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bromate/Bromide Solution, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.