missing translation for 'onlineSavingsMsg'
Learn More
Bromide bromate Standard Solution, 0.25M, Honeywell™ Product Code.: 15614780
compliance-icons

Bromide bromate Standard Solution, 0.25M, Honeywell™

Product Code. 15614780 Shop All Honeywell Fluka Products
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 15614780

Brand: Honeywell-Fluka 350071L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Volumetric, 0.25 M Br2 (0.5 N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 7758-02-3, 7758-01-2, 7732-18-5
Quantity 1 L
Molecular Formula BrK
MDL Number MFCD00011358
UN Number NONH for all modes of transport
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES [K+].[Br-]
IUPAC Name potassium bromide
Molecular Weight (g/mol) 119.00
Concentration or Composition (by Analyte or Components) 0.25M
Chemical Name or Material Bromide bromate Standard Solution
Show More Show Less
compliance-icons
Product Identifier
  • Bromide bromate Standard Solution
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : potassium bromate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bromide bromate Standard Solution, 0.25M, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.