missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromine, 99.5+%, Certified AR for Analysis, Liquid, Fisher Chemical™
Click to view available options
Quantity:
100 mL
500 mL
Specifications
Specifications
Boiling Point | 58.7°C |
Melting Point | -7.2°C |
Color | Brown-Red |
Physical Form | Liquid |
Quantity | 100 mL |
Formula Weight | 159.82g/mol |
Packaging | Plastic Coated Glass bottle |
Vapor Pressure | 230mbar at 20°C |
Density | Vapor Density: 5.51 |
Safety and Handling
Product Identifier
- Bromine
- Danger
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 1
- ACUTE AQUATIC Acute 1
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H400-Very toxic to aquatic life.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'