missing translation for 'onlineSavingsMsg'
Learn More

Bromine (Bromide-Bromate) Solution, Honeywell Fluka™

Volumetric, 0.05 M Br2 (0.1N)

Chemical Identifiers

MDL Number MFCD00143870
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15673520
View Documents Promotion Details
Honeywell-Fluka
318698-500ML
500 mL N/A
N/A
15663520
View Documents Promotion Details
Honeywell-Fluka
318698-2L
2 L N/A
N/A
Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Specifications

Chemical Identifiers

MFCD00143870

Specifications

MFCD00143870
NONH for all modes of transport
1.005g/mL at 25°C
Bromine (Bromide-Bromate) Solution
Videos
Safety and Handling

Safety and Handling

P201-P308 + P313

H350

Product Identifier
  • Bromine (bromide-bromate) solution

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1B

Hazard Statement
  • H350-May cause cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : potassium bromate, potassium bromide

SDS
Documents

Documents

Product Certifications